Learn More
Mycro 3, MedChemExpress
Product Code. 16499666
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16499666
Brand: Medchem Express HY100669
MedChemExpress Mycro 3 is a potent and selective inhibitor of Myc-associated factor X (MAX) dimerization. Mycro 3 also inhibit DNA binding of c-Myc. Mycro 3 could be used for the research of pancreatic cancer.
Mycro 3 is a potent and selective inhibitor of Myc-associated factor X (MAX) dimerization. Mycro 3 also inhibit DNA binding of c-Myc. Mycro 3 could be used for the research of pancreatic cancer.
Specifications
Chemical Name or Material Mycro 3
Physical Form Solid
CAS 944547-46-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H17ClF2N6O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (189.80 mM)
SMILES O=C(C1=C(NC(C2=NN3C(C(F)(Cl)F)=CC(C4=CC=CO4)=NC3=C2)=O)N(C5=CC=CC=C5)N=C1)OCC
Molecular Weight (g/mol) 526.88
Formula Weight 526.88
Percent Purity 99.21%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.