missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MXI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12360-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
It is now well established that Myc regulation of cell proliferation and differentiation involves a family of related transcription factors. One such factor, Max, is an obligate heterodimeric partner for Myc and can also form heterodimers with at least four related proteins designated Mad 1, Mxi1, Mad 3 and Mad 4. Like Mad 1 and Mxi1, association of Mad 3 and Mad 4 with Max results in transcriptional repression. Both Myc and the Mad proteins have short half-lives and their synthesis is tightly regulated, while Max expression is constitutive and relatively stable. Two related mammalian cDNAs have been identified and shown to encode Mad-binding proteins. Both possess sequence homology with the yeast transcription repressor Sin3 including four conserved paired amphipathic helix (PAH) domains. mSin3A and mSin3B specifically interact with the Mad proteins via their second paired amphipathic helix domain (PAH2). It has been suggested that Mad-Max heterodimers repress transcription by tethering mSin3 to DNA as corepressors.Specifications
MXI1 | |
Polyclonal | |
Unconjugated | |
MXI1 | |
bHLHc11, MAD2, Max interacting protein 1, MAX interactor 1, MXD2, MXI, MXI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17859, 25701, 4601 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O09015, P50539, P50540 | |
Mxi1 | |
MXI1 Fusion Protein Ag3024 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |