missing translation for 'onlineSavingsMsg'
Learn More

MXD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16887884 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16887884 150 μL 150µL
16897884 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16887884

Brand: Proteintech 178881AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.

Specifications

Antigen MXD1
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MXD1
Gene Accession No. P50538, Q05195
Gene Alias MAD, MAD1, MAX dimerization protein 1, Max dimerizer 1, MXD1, Protein MAD
Gene Symbols Mxd1
Host Species Rabbit
Immunogen MXD1 Fusion Protein Ag12037
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17119, 362391, 4084
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.