Learn More
MV1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
MV1 is an antagonist of IAP (inhibitor of apoptosis protein), leads to protein knockdown of HaloTag-fused proteins when combined with HaloTag ligand.
Specifications
Specifications
| Chemical Name or Material | MV1 |
| CAS | 1001600-54-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H44N4O5 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 125 mg/mL (216.74 mM) |
| SMILES | O=C(OC)[C@H](C(C1=CC=CC=C1)C2=CC=CC=C2)NC([C@H]3N(C([C@H](C4CCCCC4)NC([C@H](C)NC)=O)=O)CCC3)=O |
| Molecular Weight (g/mol) | 576.73 |
| Formula Weight | 576.73 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.