missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUTYH Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA595692

Product Code. 16588531

  • 4235.00 NOK / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MTPLVSRLSR LWAIMRKPRA AVGSGHRKQA ASQEGRQKHA KNNSQAKPSA CDGLARQPEE VVLQASVSSY HLFRDVAEVT AFRGSLLSWY DQEKRDLPWR RRAEDEMDLD RRAYAVWVSE VMLQQTQVAT VINYYTGWMQ KWPTLQDLAS ASLEEVNQLW AGLGYYSRGR RLQEGARKVV EELGGHMPRT AETLQQLLPG VGRYTAGAIA SIAFGQATGV VDGNVARVLC RVRAIGADPS STLVSQQLWG LAQQLVDPAR PGDFN.

MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MUTYH
Polyclonal
Unconjugated
MUTYH
5730495A01Rik; A/G-specific adenine DNA glycosylase; adenine DNA glycosylase; adenine-DNA glycosylase; hMYH; MGC4416; mMYH; mutY DNA glycosylase; mutY homolog; mutY homolog (E. coli); mutY homolog alpha; Mutyh; Mutyha; Mutyhalpha; Mutyhb; Mutyhbeta; Mutyhc; mutY-like protein; Myh; OTTHUMP00000009098; OTTHUMP00000009102; rMYH; RP4-534D1.2; the full-length form type A, 5' region
Rabbit
Affinity chromatography
RUO
170841, 4595
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
0.84 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R5G2, Q9UIF7
MUTYH
Recombinant fusion protein containing a sequence corresponding to amino acids 1-265 of human MUTYH (NP_0010416361).
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.