Learn More
Mutant IDH1-IN-1, MedChemExpress
Product Code. 16477917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16477917
Brand: Medchem Express HY12475
MedChemExpress Mutant IDH1-IN-1 is a mutant-selective IDH1 inhibitor with with IC50s of 4, 42, 80 and 143 nM against mutant IDH1 R132C/R132C, IDH1 R132H/R132H, IDH1 R132H/WT and wild type IDH1, respectively.
Mutant IDH1-IN-1 is a mutant-selective IDH1 inhibitor with with IC50s of 4, 42, 80 and 143 nM against mutant IDH1 R132C/R132C, IDH1 R132H/R132H, IDH1 R132H/WT and wild type IDH1, respectively.
Specifications
Chemical Name or Material Mutant IDH1-IN-1
CAS 1355326-21-4
Color White
Purity Grade Notes Research
Molecular Formula C30H31FN4O2
Quantity 5 mg
Solubility Information DMSO : ≥ 45 mg/mL (90.25 mM)
SMILES O=C(C(C1=C(C)C=CC=C1)N(C(CN2C=NC3=C2C=CC=C3)=O)C4=CC(F)=CC=C4)NC5CCCCC5
Molecular Weight (g/mol) 498.59
Formula Weight 498.59
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.