Learn More
BD Biosciences™ Multitest CD3/CD8/CD45/CD4 (CE/IVD)
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
Description
- Four-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T (CD3+), natural killer (NK) (CD3–CD16+CD56+), and B (CD3–CD19+) lymphocytes in erythrocyte-lysed whole blood.
- When used with BD Trucount™ tubes, absolute counts of these populations can be enumerated from a single tube.
- BD Multitest CD3/CD16+CD56/CD45/CD19 and BD Trucount tubes can be used with the BD FACS™ Loader, BD FACSVia™ Loader, and BD FACS™ Universal Loader
Specifications
Specifications
| Content And Storage | Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. |
| Includes | Trucount™ Tubes |
| No. of Tests | 50 Tests |
| Quantity | 50 Tests |
| Regulatory Status | CE/IVD |
| Product Type | Immunofluorescence Flow Cytometry Test Reagents |