Learn More
ZVG™ Multisoft™ Air+ Wipes
Product Code. 17883665
Click to view available options
Dimensions (L x W):
27 x 32 cm
32 x 34 cm
Unit Size:
Pack of 1000
Pack of 500
Product Code. 17883665
Brand: ZVG™ 1010400
Only null left
Add to basket
Add to basket
These lint-free, highly absorbent, wipes are made of soft embossed white cellulose.
Sensitive surfaces require gentle treatment. Multisoft™ extra soft, low-lint specialty cloths are an ideal choice for care and polishing tasks. They efficiently absorb liquids, such as polishing wax, and evenly release it without compromising their strength. In the zetPutz™ range, you will find additional low-lint, highly absorbent 1- to 4-ply cleaning and polishing cloths for various cleaning purposes.
  • Airlaid material
  • Highly absorbent
  • Embossed
  • Latex- and silicone-free
  • Solvent-resistant
Specifications
Product Type Wipe
Length (English) 12.6 in.
Length (Metric) 32 cm
Width (English) 13.4 in.
Width (Metric) 34 cm
Material Fleece
Color White
Disposable Single-use
Dimensions (L x W) 32 x 34 cm
Packaging Quantity 500 Cloths