Learn More
Brady™ Multipurpose Vinyl Label Tape for M6/M7 Printers
With availability in 12 colors, colored vinyl labels are ideal for product, component, rating plate and general identification where various color options is required.
Brand:  Brady™ 174318
979.07 NOK valid until 2026-10-31
BEST PRICE on Promo! Use promo code "28026" to get your promotional price.
 View more versions of this product
Product Code. 17888834
  • 1340.00 NOK / Pack of 1
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 22-10-2026
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty, and harsh environment surfaces, and can be applied in cold temperatures
  • Designed for use in ambient conditions with limited solvent exposure
  • RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-439 vinyl material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Continuous
Single-side Printable
Label
Barcode Labeling, Equipment Labeling, Electrical Labeling, General Identification, Inventory and Inspection Labeling
No
Rectangle
-70°C to +80°C
0.107 mm
50 ft.
1.9 in.
M6C-1900-439
ASTM D 1000, ASTM D 2979, ASTM G 155
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-94°F to +176°F
0.0042 in.
50 ft. x 1.9 in.
15.24 m
48.26 mm
Certificates
Promotions
Promotions
Promo
Expires: 2026-10-31
Support a Safer, More Efficient Laboratory
Promo Code: 28026
Save up to 66% on trusted facility safety and maintenance solutions.
Applying Promo and Adding...
Promo
Expires: 2026-10-31
Safety Starts Here: Save Up to 60%
Promo Code: 28008
Make safety a priority while making your budget go further. For a limited time, save up to 60% across a wide selection of safety essentials for your laboratory and workplace.
Applying Promo and Adding...