Learn More
Benchmark Scientific™ Multipurpose Block
Click to view available options
Benchmark Scientific™ Multipurpose Block
Description
- Multipurpose block with 4 x 2.6.1 in. chamber
- Ideal for use with metal beads and odd shaped containers
- Protective shield protects users from burns
- Attached lid helps to maintain temperature
- Fits MultiTherm H and HC Models
Specifications
Autoclavable | Not Autoclavable |
Chamber Material | Aluminum |
Holds | Mulitpurpose Blocks |
Material | Aluminum |
No. of Places | 1 |
Type | Block |