Learn More
Benchmark Scientific™ Multipurpose Block
Catalog Number 17974402
Click to view available options
Benchmark Scientific™ Multipurpose Block
This multipurpose block has a 4 x 2.6 x 1 in. interior for holding metal beads, liquids or other containers. Made of aluminum with protective shield and attached lid. Fits Benchmark MultiTherm.
  • Multipurpose block with 4 x 2.6.1 in. chamber
  • Ideal for use with metal beads and odd shaped containers
  • Protective shield protects users from burns
  • Attached lid helps to maintain temperature
  • Fits MultiTherm H and HC Models
Autoclavable Not Autoclavable
Chamber Material Aluminum
Holds Mulitpurpose Blocks
Material Aluminum
No. of Places 1
Type Block