Learn More
Proteintech MultiPro™ Human Fixed Cell Immune Profiling Antibody Cocktail
Shop All Proteintech ProductsDescription
MultiPro™ Human Fixed Cell Immune Profiling Antibody Cocktail
Specifications
Specifications
| Quantity | 4 Tests |
| Primary or Secondary | Primary |
| Form | Liquid |