Learn More
Shamrock™ Multiple Tape Dispenser for 2160 in. Rolls
Click to view available options
For Use With (Equipment):
Label tape on rolls
Unit Size:
Each
Description
- Holds 8 rolls
- With 5 dividers
Specifications
Specifications
| Color | White |
| For Use With (Application) | Storage and dispensing |
| For Use With (Equipment) | Label tape on rolls |