Learn More
WTW™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meter
Solve multiple measuring tasks at once and save time and costs. WTW7™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meters feature three universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
Brand: WTW™ 2FD570
26202.82 NOK valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Additional Details : Weight : 2.50000kg
Description
- Three channels, four parameters
- Three measuring channels, freely combinable for the same or different parameters
- For simultaneous multi-parameter measurement without compromise
- Also use the wireless modules together with the new IDS plug head sensors to be independent from cables and measure conveniently at difficult-to-access locations
- Allows the operation with the digital multi-parameter probes MPP 910 IDS and MPP 930 IDS
- Brilliant color graphic display and two USB interfaces
- The batteries can be charged directly in the device
- Archiving user administration can also be used alternatively to mark measuring points
- 10,000 data sets include up to three main parameters, three temperature values as well as the water depth if using the depth probe
Specifications
Multi 3630 IDS SET F | |
3 | |
Basic meter unit only with power supply and USB cable | |
Multi 3630 IDS | |
Multi-parameter portable meter |
Yes | |
Multi 3630 IDS SET KS2 | |
Manual: 500 records ; Automatic: 10,000 records | |
Wide range power supply, 4 x 1.2 V NiMH battery or via Mini USB-B |