Learn More
Velp Scientifica™ MultiAluBlock™ Block with Eight Positions (17 x 26 mm)
Product Code. 15865201 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
AREX-6 Hotplate Stirrers
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15865201 AREX-6 Hotplate Stirrers Each
1 options
Product Code. 15865201
Brand: Velp Scientifica™ A00000328
Only null left
Add to basket
Add to basket
For Use With: AREX-6 Hotplate Stirrers
  • Used to perform numerous reactions reducing time and bench space
  • For use with Velp Series 6 Hotplate Stirrers
  • Interchangable with eachother
Specifications
Diameter (Metric) 17 mm
For Use With (Equipment) AREX-6 Hotplate Stirrers
Height (Metric) 26 mm
Holds 8 Positions
Type Block
Width (Metric) 26 mm