Learn More
Sampling Systems™ Multi Dipper 2
Product Code. 15839341 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
1000 mm
1500 mm
2000 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
15839341 1500 mm Each
15829341 1000 mm Each
15849341 2000 mm Each
3 options
Product Code. 15839341
Brand: Sampling Systems™ 5045A1500
Only null left
Add to basket
Add to basket
Maintain a closed sample collection bottle until the operator uses the control chain to remove the stoppers. Sampling Systems™ Multi Dipper 2 is the ideal solution for taking liquid samples from different levels.
  • Sampler frame, control chains, plugs and plug shackles are constructed of Stainless Steel
  • Sampler bottle are constructed of High Density Polyethylene (HDPE)
  • Ideal solution for taking liquid samples from different levels
  • The sample collection bottles remain closed until the operator uses the control chain to remove the stoppers
Specifications
Product Type Dipper
Material Stainless Steel (316L)
Includes 3 HDPE Sampling Bottles
Capacity (Metric) 3 x 250 mL
For Use With (Application) Liquid Sampling
Length (Metric) 1500 mm