Learn More
Shamrock™ Multi-Purpose Labels, 25 x 50mm
Click to view available options
:
25 x 50mm
Description
- Economic with a wide array of uses
- Permanent marking
- Universal marking with a ballpoint pen, paper pencil and felt marker
- Labels pre-cut on roll
Specifications
Specifications
| Dimensions (LxW) | 25 x 50 mm |