Learn More
Nabertherm™ Muffle Furnaces with Brick Insulation and Lift Door
Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
15 L
Unit Size:
Each
Description
- Heating from two sides
- Heating elements on support tubes ensure free heat radiation and a long service life
- Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
- Housing made of sheets of textured stainless steel
- Dual shell housing for low external temperatures and stability
- Lift door (LT) with hot surface facing away from operator
- Adjustable air inlet in the furnace door
- Exhaust air outlet in rear wall of furnace
- Solid state relays provide for low-noise operation
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
- Chimney, chimney with fan or catalytic converter
- Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load
- Protective gas connection to purge with non-flammable protective or reaction gases
- Manual or automatic gas supply system
Specifications
Specifications
| Controller Type | C450 |
| Capacity (Metric) | 15 L |
| Power Consumption | 3.5 kw |
| Dimensions (D x W x H) Interior | 340 x 260 x 170 mm |
| Width (Metric) | 530 mm |
| Height (Metric) | 630 mm |
| Depth (Metric) | 625 mm |