Learn More
Cole-Parmer™ Mucksucker Drum and Tank Sampler
Product Code. 12776047 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (English):
3.5 ft.
Unit Size:
Each
Product Code. Length (English) unitSize
12776047 3.5 ft. Each
1 options
Product Code. 12776047
Brand: Cole-Parmer™ 9920020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Denna artikel kan inte returneras. Se returpolicy
High-suction syringe action is ideal for viscous fluids
  • Drum and tank sampler.
  • 3.5 ft. long.
  • Efficiently draws samples that are close to the bottom of the drum or tank.
  • Comes with both a push/seal bottom and a coring bottom.
  • Coring bottom features a wide mouth for handling sludge samples.
Specifikationer
Product Type Drum and Tank Sampler
Length (English) 3.5 ft.
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.