Learn More
Invitrogen™ MUC5B Monoclonal Antibody (5B19-2E)

Description
Both IHC and WB applications require the following deglycosylation protocols. Immunohistochemistry 1. Prepare the following stock solutions: a. 200 mM NaIO4 (8.556 g/200 mL water) b. Sodium-Acetate buffer, pH 4.5: Dissolve NaCl (final concentration: 0.33M) into 0.1M glacial acetic acid and adjust to pH 4.5 using 1 N NaOH. c. Sodium-thiosulfate solution: Dissolve Na2S2O3 (0.133M), NaI (0.033M), and NaHCO3 (0.033M) in water and adjust to pH 7.6 using 1M HCl. Keep solution at 4°C. 2. After deparaffinization, rehydration, and peroxidase quenching steps, mix a 1:1 solution of a and b (in step 1), add to each slide, and incubate overnight at 4°C. 3. Treat tissue with solution c for 30 minutes at 4°C. 4. Proceed with normal immunostaining protocol (blocking and antibody incubation steps). Western blotting 1. Treat transblotted membrane with 10 mM NaIO4 in 0.1M sodium acetate/5 mM EDTA, pH 5.5 overnight at 4°C in the dark. 2. After washing off the solution, incubate the membrane in a solution containing 30 mM Na2S2O3, 7.5 mM NaI, and 7.5 mM NaHCO3, pH 7.2 for 1 hour at 4°C in the dark. 3. Proceed with normal Western blot protocol (wash and blocking steps).
Specifications
Specifications
| Antigen | MUC5B |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5B19-2E |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | MUC5B |
| Gene Accession No. | Q9HC84 |
| Gene Alias | Cervical mucin; cervical mucin MUC5B; high molecular weight salivary mucin MG1; MG1; MUC5; MUC5B; MUC-5B; MUC9; mucin 5, subtype B, tracheobronchial; mucin 5B, oligomeric mucus/gel-forming; Mucin-5 subtype B, tracheobronchial; Mucin-5B; sublingual gland mucin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.