Learn More
Invitrogen™ MUC21 Polyclonal Antibody
GREENER_CHOICE
Product Code. 15730332 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
15730332 100 μL 100µL
1 options
Product Code. 15730332
Brand: Invitrogen™ PA562536
Rabbit Polyclonal Antibody
Immunogen sequence: CVRNSLSLRN TFNTAVYHPH GLNHGLGPGP GGNHGAPHRP RWSPNWFWRR PVSSIAMEMS GRNS Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 27%.
TRUSTED_SUSTAINABILITY
Specifications
Antigen MUC21
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MUC21
Gene Accession No. Q5SSG8
Gene Alias C6orf205; Epiglycanin; KMQK697; MUC21; MUC-21; mucin 21, cell surface associated; mucin-21; UNQ697/PRO1342
Gene Symbols MUC21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MUC21. Recombinant protein control fragment (Product #RP-99560).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 394263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG