missing translation for 'onlineSavingsMsg'
Learn More

MTRF1 Monoclonal Antibody (OTI3A11), TrueMAB™, OriGene

Product Code. 17597734
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17597734 30 μL 30µL
17577734 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17597734

Brand: Origene Technologies TA813066S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MTRF1 Monoclonal antibody specifically detects MTRF1 in Human samples. It is validated for Western Blot

Specifications

Antigen MTRF1
Applications Western Blot
Classification Monoclonal
Clone OTI3A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MTRF1
Gene Accession No. O75570
Gene Alias A830062K05Rik; mitochondrial translational release factor 1; mitochontrial peptide chain release factor 1; MRF1; MRF-1; Mtrf1; MtRF-1; MTTRF1; Peptide chain release factor 1, mitochondrial; RF1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 240-445 of human MTRF1 (NP_004285) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9617
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.