Learn More
MTR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25896-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16894445
  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
Description
MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G.
Specifications
Specifications
MTR
Polyclonal
Unconjugated
MTR
cblG, Methionine synthase, MS, MTR
Rabbit
Antigen Affinity Chromatography
RUO
238505, 4548
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6H5Y3, Q99707
MTR
MTR Fusion Protein Ag23156
150 μL
Primary
Human, Mouse