Learn More
MTR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25896-1-AP-20UL
This item is not returnable.
View return policy
Description
MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G.Specifications
| MTR | |
| Polyclonal | |
| Unconjugated | |
| MTR | |
| cblG, Methionine synthase, MS, MTR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 238505, 4548 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6H5Y3, Q99707 | |
| MTR | |
| MTR Fusion Protein Ag23156 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |