Learn More
mTOR Mouse anti-Human, Unconjugated, Clone: 1G11A3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66888-1-IG-150UL
 View more versions of this product
Product Code. 16899155
  • 4836.10 NOK / 150µL
Description
The mTOR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human mTOR. This antibody recognizes human, mouse, rat antigen. The mTOR antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
mTOR
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
1G11A3
Unconjugated
150 μL
2475
IgG2a