Learn More
mTOR Mouse anti-Human, Unconjugated, Clone: 1G11A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66888-1-IG-150UL
Description
The mTOR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human mTOR. This antibody recognizes human, mouse, rat antigen. The mTOR antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| mTOR | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1G11A3 | |
| Unconjugated | |
| 150 μL | |
| 2475 | |
| IgG2a |