missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12299-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the myotubularin-related family and is part of the MTMR6 subgroup. Family members are dual-specificity phosphatases and the encoded protein contains a phosphoinositide-binding domain (PID) and a SET-interacting domain (SID). Defects in other family members have been found in myotubular myopathic diseases.Specifications
| MTMR8 | |
| Polyclonal | |
| Unconjugated | |
| MTMR8 | |
| MTMR8, myotubularin related protein 8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55613 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96EF0 | |
| MTMR8 | |
| MTMR8 Fusion Protein Ag2944 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction