missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25990-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the myotubularin family of tyrosine/dual-specificity phosphatases. The encoded protein is characterized by four distinct domains that are conserved among all members of the myotubularin family: the glucosyltransferase, Rab-like GTPase activator and myotubularins domain, the Rac-induced recruitment domain, the protein tyrosine phosphatases and dual-specificity phosphatases domain and the suppressor of variegation 3-9, enhancer-of-zeste, and trithorax interaction domain. This protein dephosphorylates the target substrates phosphatidylinositol 3-phosphate and inositol 1, 3-bisphosphate. A pseudogene of this gene is found on chromosome 5.Specifications
MTMR7 | |
Polyclonal | |
Unconjugated | |
MTMR7 | |
MTMR7, myotubularin related protein 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306490, 54384, 9108 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y216, Q9Z2C9 | |
Mtmr7 | |
MTMR7 Fusion Protein Ag23170 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |