missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14372-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
MTMR2 | |
Polyclonal | |
Unconjugated | |
MTMR2 | |
CMT4B, CMT4B1, KIAA1073, MTMR2, myotubularin related protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315422, 77116, 8898 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13614, Q9Z2D1 | |
Mtmr2 | |
MTMR2 Fusion Protein Ag5745 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |