missing translation for 'onlineSavingsMsg'
Learn More

MTMR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14372-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892984

  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

MTMR2
Polyclonal
Unconjugated
MTMR2
CMT4B, CMT4B1, KIAA1073, MTMR2, myotubularin related protein 2
Rabbit
Antigen Affinity Chromatography
RUO
315422, 77116, 8898
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13614, Q9Z2D1
Mtmr2
MTMR2 Fusion Protein Ag5745
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers