missing translation for 'onlineSavingsMsg'
Learn More

MTF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16833965 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16833965 150 μL 150µL
16843965 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16833965

Brand: Proteintech 253831AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).

Specifications

Antigen MTF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MTF1
Gene Accession No. Q07243, Q14872
Gene Alias MTF 1, MTF1, Transcription factor MTF 1, ZRF
Gene Symbols Mtf1
Host Species Rabbit
Immunogen MTF1 Fusion Protein Ag21848
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17764, 4520
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.