missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25383-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).Specifications
| MTF1 | |
| Polyclonal | |
| Unconjugated | |
| MTF1 | |
| MTF 1, MTF1, Transcription factor MTF 1, ZRF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17764, 4520 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q07243, Q14872 | |
| Mtf1 | |
| MTF1 Fusion Protein Ag21848 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction