Learn More
MTCO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55070-1-AP-20UL
 View more versions of this product
Product Code. 16856515
  • 1710.33 NOK / 20µL
Description
The antibody is specific to COX2.
Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.
Specifications
Specifications
MTCO2
Polyclonal
Unconjugated
COX2
COII, COX2, COXII, cytochrome c oxidase II, Cytochrome c oxidase subunit 2, MT CO2, MTCO2
Rabbit
Antigen Affinity Chromatography
RUO
17709, 26198, 4513
-20°C
Liquid
Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00403, P00405
COX2
Peptide
20 μL
Primary
Rat, Mouse, Human