Learn More
Abnova™ MT1F (Human) Recombinant Protein
Product Code. 16127981
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16127981 25 μg 25µg
16117981 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16127981 Supplier Abnova™ Supplier No. H00004494P01.25ug
Human MT1F full-length ORF ( AAH29453, 1 a.a. - 61 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MDPNCSCAAGVSCTCAGSCKCKECKCTSCKKSCCSCCPVGCSKCAQGCVCKGASEKCSCCD
Specifications
Accession Number AAH29453
Gene ID (Entrez) 4494
Name metallothionein 1F
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias MGC32732, MT1
Gene Symbol MT1F
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer