Learn More
HUBER PTFE Hose K-260-1-M24x1,5
Product Code. 10364425
Click to view available options
Length (Metric):
1000 mm
Unit Size:
Each
Product Code. 10364425
Brand: HUBER 9325
Specifications
Connection Type M24 x 1.5 Thread
Material PTFE
Length (Metric) 1000 mm
Includes with Braided Metal Reinforcement
Quantity 1 Ea.
Weight (Metric) 1.3 kg
Type Hose