Learn More
HUBER PTFE Hose K-260-1,5-M24x1,5
Brand: HUBER 9326
Additional Details : Weight : 0.80000kg
Specifications
M24 x 1.5 Thread | |
1500 mm | |
1 Ea. | |
Hose |
PTFE | |
with Braided Metal Reinforcement | |
1.8 kg |