Learn More
MSX-122, MedChemExpress
Product Code. 16488477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16488477
Brand: Medchem Express HY13696
MedChemExpress MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ∼10 nM. MSX-122 has anti-inflammatory and anti-metastatic activity.
MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ∼10 nM. MSX-122 has anti-inflammatory and anti-metastatic activity.
Specifications
Chemical Name or Material MSX-122
Physical Form Solid
CAS 897657-95-3
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H16N6
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 4 mg/mL (13.68 mM; Need ultrasonic)
SMILES C1(CNC2=NC=CC=N2)=CC=C(CNC3=NC=CC=N3)C=C1
Molecular Weight (g/mol) 292.34
Formula Weight 292.34
Percent Purity 96.56%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.