Learn More
MSTO1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28400-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The MSTO1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MSTO1. This antibody recognizes human antigen. The MSTO1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
MSTO1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
55154 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human |