Learn More
MSTO1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28400-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16876125
  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
Description
The MSTO1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MSTO1. This antibody recognizes human antigen. The MSTO1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MSTO1
Polyclonal
Unconjugated
150 μL
55154
IgG
Western Blot, Immunohistochemistry, ELISA
300 μg/mL