missing translation for 'onlineSavingsMsg'
Learn More

MSPR/Ron Beta Antibody (narnatumab) [Janelia Fluor™ 525], R&D Systems™

Product Code. 30171572
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171572 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171572 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328640JF525
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Human Monoclonal Antibody

MSPR/Ron Beta Monoclonal specifically detects MSPR/Ron Beta in Human samples. It is validated for ELISA, Flow Cytometry, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSPR/Ron Beta
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone narnatumab
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CD136, CDw136, Macrophage-stimulating protein receptor beta chain, MSP receptor, NPCA3, p185-Ron, Protein-tyrosine kinase 8, PTK8, Ron
Host Species Human
Immunogen MSPR/ RON/ CD136
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4486
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.