missing translation for 'onlineSavingsMsg'
Learn More

MSH6 Mouse anti-Human, Clone: 2E10B2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66172-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847805

  • 1415.13 NOK / 20µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced maligt transformation and hence the prevention of skin cancer.
Specifications

Specifications

MSH6
Monoclonal
1.03 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52701
MSH6
MSH6 Fusion Protein Ag12645
20 μL
Primary
Human
Antibody
IgG2a
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
2E10B2
Unconjugated
MSH6
G/T mismatch binding protein, GTBP, GTMBP, hMSH6, HNPCC5, HSAP, MSH6, MutS alpha 160 kDa subunit, mutS homolog 6 (E. coli), p160
Mouse
Protein A
RUO
2956
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers