missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15520-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Plays a role in neurogenesis and neuronal migration By similarity. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.Specifications
MSH2 | |
Polyclonal | |
Unconjugated | |
MSH2 | |
COCA1, FCC1, hMSH2, HNPCC, HNPCC1, LCFS2, MSH2, MutS protein homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17685, 4436, 81709 | |
-20°C | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43246, P43247, P54275 | |
Msh2 | |
MSH2 Fusion Protein Ag7835 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |