missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Monoclonal Antibody (OTI10F7), TrueMAB™, OriGene
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
MSH2 Monoclonal antibody specifically detects MSH2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MSH2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI10F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Msh2 |
| Gene Accession No. | P43246, P43247, P54275 |
| Gene Alias | AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?