Learn More
STEMCELL Technologies MSC Characterization Antibody Panel
Product Code. 17177571
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
17177571 1 Kit 1 set
1 options
Product Code. 17177571
Brand: STEMCELL Technologies 1000354
Multi-color flow cytometry kit for characterization of human mesenchymal stromal cells
  • The MSC Characterization Antibody Panel is designed for the phenotypic analysis of human mesenchymal stromal cells (MSCs)
  • This kit contains three fluorochrome-conjugated antibodies for the identification of human MSCs, and a fluorochrome-conjugated antibody for the identification of non-MSCs
  • When measured by flow cytometry, ≥ 95% of the MSC population must express CD73, CD90, and CD105, and these cells must lack expression (≤ 2% positive) of CD45
  • These reagents provide single-step labeling for the verification of human MSCs
  • The MSC Characterization Antibody Panel can be used to identify human MSCs derived from various tissues such as bone marrow MSCs, pluripotent stem cell-derived mesenchymal progenitor cells, adipose-derived MSCs, dental pulp-derived MSCs, and umbilical cord-derived MSCs
Specifications
Applications Flow Cytometry
Classification Monoclonal
Quantity 1 Kit
Cell Type Mesenchymal Stromal Cells