missing translation for 'onlineSavingsMsg'
Learn More

MS4A8B Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30499984 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499984 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30499984

Brand: Novus Biologicals NBP335124IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MS4A8B Polyclonal antibody specifically detects MS4A8B in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A8B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 4SPAN4, Four-span transmembrane protein 4, membrane-spanning 4-domains subfamily A member 8B, membrane-spanning 4-domains, subfamily A, member 8B, MS4A4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MS4A8B (NP_113645.1).,, Sequence:, SISFYGGFPFWGGLWFIISGSLSVAAENQPYSYCLLSGSLGLNIVSAICSAVGVILFITDLSIPHPYAYPDYYPYAWGVNPGMAISGVLLVFCLLEFGIAC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83661
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.