missing translation for 'onlineSavingsMsg'
Learn More

MRT68921 dihydrochloride, MedChemExpress

Product Code. 16425477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425477

Brand: Medchem Express HY100006A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRT68921 dihydrochloride is a potent inhibitor of ULK1 and ULK2, with IC50 values of 2.9 nM and 1.1 nM, respectively.

MRT68921 dihydrochloride is a potent inhibitor of ULK1 and ULK2, with IC50 values of 2.9 nM and 1.1 nM, respectively.

Specifications

Chemical Name or Material MRT68921 dihydrochloride
CAS 2080306-21-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H36Cl2N6O
Quantity 5 mg
Solubility Information H2O : 31.25 mg/mL (61.58 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (16.41 mM; ultrasonic and warming and heat to 60°C)
SMILES [H]Cl.O=C(C1CCC1)NCCCNC2=NC(NC3=CC4=C(CN(C)CC4)C=C3)=NC=C2C5CC5.[H]Cl
Molecular Weight (g/mol) 507.5
Formula Weight 507.5
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.