missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The MRRF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRRF. This antibody recognizes human antigen. The MRRF antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | MRRF |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1G5C3 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 92399 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?