missing translation for 'onlineSavingsMsg'
Learn More

MRPS25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16854384 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16854384 20 μL 20µL
16844384 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16854384

Brand: Proteintech 152771AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody specifically recognizes the 18 kDa MRPS25 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 4.

Specifications

Antigen MRPS25
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MRPS25
Gene Accession No. P82663, Q4QR80, Q9D125
Gene Alias MRP S25, MRPS25, RPMS25, S25mt
Gene Symbols Mrps25
Host Species Rabbit
Immunogen MRPS25 Fusion Protein Ag7395
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297459, 64432, 64658
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.