Learn More
MRPS16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16735-1-AP-20UL
 View more versions of this product
Product Code. 16856464
  • 1710.33 NOK / 20µL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S16P family. The encoded protein is one of the most highly conserved ribosomal proteins between mammalian and yeast mitochondria. Three pseudogenes (located at 8q21.3, 20q13.32, 22q12-q13.1) for this gene have been described.
Specifications
Specifications
MRPS16
Polyclonal
Unconjugated
MRPS16
CGI 132, COXPD2, MRP S16, MRPS16, RPMS16, S16mt
Rabbit
Antigen Affinity Chromatography
RUO
51021, 66242, 688912
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CPX7, Q9Y3D3
MRPS16
MRPS16 Fusion Protein Ag9749
20 μL
Primary
Human, Rat, Mouse