missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL50 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MRPL50.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a putative 39S subunit protein and belongs to the L47P ribosomal protein family. Pseudogenes corresponding to this gene are found on chromosomes 2p, 2q, 5p, and 10q. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigen | MRPL50 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRPL50 |
| Gene Accession No. | Q8N5N7 |
| Gene Alias | 39S ribosomal protein L50, mitochondrial; L50mt; mitochondrial 39S ribosomal protein L50; Mitochondrial large ribosomal subunit protein mL50; mitochondrial ribosomal protein L50; MRPL50; MRP-L50 |
| Gene Symbols | MRPL50 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?