missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538961
Additional Details : Weight : 0.00900kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L22 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 4q. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].Specifications
MRPL22 | |
Polyclonal | |
Unconjugated | |
MRPL22 | |
MRPL25, RPML25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216767, 287302, 29093 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NWU5, Q8BU88, P0C2C0 | |
MRPL22 | |
A synthetic peptide derived from the internal region of human MRPL22 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |