missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MR1 Antibody (MR1/7577) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26943JF669
5010.75 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MR1 Monoclonal antibody specifically detects MR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MR1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein | |
| Recombinant fragment (around aa50-250) of human MR1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 3140 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| MR1/7577 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction