missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPRIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20040-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of isoform 2 and isoform 3 of MPRIP.
Targets myosin phosphatase to the actin cytoskeleton. Required for the regulation of the actin cytoskeleton by RhoA and ROCK1. Depletion leads to an increased number of stress fibers in smooth muscle cells through stabilization of actin fibers by phosphorylated myosin. Overexpression of MRIP as well as its F-actin-binding region leads to disassembly of stress fibers in neuronal cells.Specifications
| MPRIP | |
| Polyclonal | |
| Unconjugated | |
| MPRIP | |
| KIAA0864, M RIP, MPRIP, MRIP, p116Rip, Rho interacting protein 3, RHOIP3, RIP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116504, 23164, 26936 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97434, Q6WCQ1, Q9ERE6 | |
| MPRIP | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction